Merck
Z380229
Jars Nalgene® style 2118
capacity 60 mL
Synonym(e):
laboratory jars, nalgene jars
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
Materialien
polypropylene bottle
polypropylene cap
round translucent bottle
Verbindung
Joints threaded neck (53)
Beschreibung
Autoclavable with closure completely unscrewed
Leistungsmerkmale
with cap
Verpackung
pkg of 12
Hersteller/Markenname
Nalgene® 2118-0002
Kapazität
60 mL
Allgemeine Beschreibung
Translucent with excellent chemical resistance.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)