Über diesen Artikel
NACRES:
NB.43
UNSPSC Code:
41121800
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
amber high-density polyethylene
packaging
case of 72 ea
manufacturer/tradename
Nalgene DS2185-0004
capacity
125 mL
volume
125 mL
General description
Perform short-term general purpose laboratory and field applications with Thermo Scientific Nalgene Wide-Mouth HDPE Economy Bottles with Closure. These light-duty translucent bottles are a lightweight version of the original Nalgene bottle, bulk-packed for ultimate economy. Thinner-walled and more flexible than the Nalgene Lab Quality Bottles, they feature a wide mouth for easy filling and offer excellent chemical resistance to most acids, bases and alcohols.
Also available in opaque amber color which reduces UV light transmissions to protect light-sensitive liquids.
Also available in opaque amber color which reduces UV light transmissions to protect light-sensitive liquids.
Features and Benefits
- Linerless polypropylene caps ensure leakproof† performance without a liner that can wrinkle, cause leaks and contaminate your solutions
- Recommended for sample collection and storage of daily working solutions
- Participates in My Green Lab′s ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
- Leakproof† when used with Nalgene closures
- Bottle and carton are readily recyclable in most communities
- Manufactured from non-toxic materials
- Opaque Bottles can be used in freezer to -100°C (-148°F)
- Opaque bottles meet U.S. Pharmacopoeia 23 requirements for maximum light transmission.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Still not finding the right product?
Explore all of our products under Nalgene® economy wide-mouth bottle
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?