Merck
TMO312002-9125
Nalgene® diagnostic bottle
natural high-density polyethylene, volume 4 mL, case of 2000 ea
Synonym(e):
PPCO bottles, small volume bottle with closure
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Materialien
natural high-density polyethylene
Verpackung
case of 2000 ea
Hersteller/Markenname
Nalgene 312002-9125
Kapazität
4 mL
Volumen
4 mL
Allgemeine Beschreibung
Sample, store and ship small volume, valuable reagents. Thermo Scientific Nalgene Natural PPCO Diagnostic Bottles with Closure are durable, autoclavable and used in test kits and many small-volume applications. These natural, translucent polypropylene copolymer bottles feature uniform walls resistant to splitting or puncturing. Bottles include linerless polypropylene closures that ensure leakproof† performance without a liner that can wrinkle, cause leaks and contaminate your solutions.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)