PURGR
Pur-A-Lyzer™ Floating Racks for Dialysis
Mega
Synonym(e):
Dialyzers, High through put dialysis tubing
Verpackung
pack of 1 ea
Verwandte Kategorien
Allgemeine Beschreibung
Floating rack can process up to 1 Mega Pur-A-Lyzer Dialysis Tube in a single beaker.
Rechtliche Hinweise
Pur-A-Lyzer is a trademark of Sigma-Aldrich Co. LLC