Merck
MTCR1070
[MTC"] Bio Tube Rack
to hold, 30 x 15 mL and 20 x 50 mL tubes, orange, pack of 5 ea
Synonym(e):
laboratory rack, stacking western blot box, tube holder
About This Item
UNSPSC-Code:
41122804
NACRES:
NB.12
Materialien
orange
polypropylene
Leistungsmerkmale
to hold 30 x 15 mL and 20 x 50 mL tubes
Verpackung
pack of 5 ea
Hersteller/Markenname
MTC Bio R1070
Allgemeine Beschreibung
Rack for 30x15 & 20x50ml (17x20x6cm), Orange, 5/pk
Leistungsmerkmale und Vorteile
  • Polypropylene, withstand -80°C
  • Compact and space efficient
  • Autoclavable
Rechtliche Hinweise
MTC is a trademark of MTC Bio