Merck
HS120623
MagFuge High Speed Centrifuge Rotor
to hold, 6 x 5 mL tubes
About This Item
UNSPSC-Code:
41103909
NACRES:
NB.78
Leistungsmerkmale
to hold 6 x 5 mL tubes
Verpackung
pkg of 1 ea
Hersteller/Markenname
Heathrow Scientific 120623
Parameter
12,500 rpm speed (max.)
9,783 × g max. RCF
Kompatibilität
Heathrow Scientific HS10050
Allgemeine Beschreibung
High speed centrifuge rotor for MagFuge.
Rechtliche Hinweise
MagFuge is a trademark of Heathrow Scientific