Merck
GE28-4064-10
Sigma-Aldrich
Tricorn 5/100 Column
Synonym(e):
5/100 Tricorn Column, Tricorn Column
About This Item
UNSPSC-Code:
41115700
NACRES:
SB.52
Materialien
glass column
Verpackung
1 ea of
Allgemeine Beschreibung
Tricorn 5 and 10 Coarse Filter Kit is used for large volume/repeated loading of clarified sample to reduce the risk of clogging in combination with capture media. Both filter and coarse filter kits include top and bottom adapters, bottom filters, and EPDM O-rings, five of each.
Tricorn Packing Equipment includes the Packing Equipment 5/50 (5-mm connector and 50-mm glass tube) or 10/100 (10-mm connector and 100-mm glass tube), EPDM O-rings and bottom unit, cap, and stop plug.
Tricorn columns include a glass tube, adapter unit, end cap, filter kit, two stop plugs, two fingertight connectors, two M6 connectors, adapter lock, and filter holder.
Leistungsmerkmale und Vorteile
Broad range of column dimensions suitable for many different purification needs.
Glass tubes coated with plastic film prevent harm if the column is dropped or operated outside its specification.
Locking ring prevents accidental compression of media bed.
Separate packing connector allows simple column packing for a broad range of media.
The modern integrated, snap-on design results in a column that is easy to assembly, operate, and maintain.
Tricorn empty columns are designed for high performance separations on small and medium scale. Pressure stability up to 100 bar and durable column material ensure reliable and reproducible operation as well as compatibility with the majority of the commercial lab media.
Empfohlene Produkte
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)