Merck
GBL664273
Grace Bio-Labs CultureWell silicone isolator sheet material
thickness 1.6 mm, (No adhesive)
Synonym(e):
silicone sheet
About This Item
UNSPSC-Code:
41122600
NACRES:
NB.22
Materialien
clear silicon
Leistungsmerkmale
(No adhesive)
Verpackung
pack of 5 ea
Hersteller/Markenname
Grace Bio-Labs 664273
L × B
180 mm × 130 mm
Dicke
1.6 mm
Allgemeine Beschreibung
CultureWell clear silicone sheet material allows researchers to create their own removable hydrophobic barriers to isolate specimens. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides.
Rechtliche Hinweise
Culturewell is a trademark of Grace Bio-Labs, Inc.
Die Dokumentenbibliothek aufrufen
Paula G Miller et al.
Biotechnology and bioengineering, 117(2), 486-497 (2019-10-15)
Efficient and economical delivery of pharmaceuticals to patients is critical for effective therapy. Here we describe a multiorgan (lung, liver, and breast cancer) microphysiological system ("Body-on-a-Chip") designed to mimic both inhalation therapy and/or intravenous therapy using curcumin as a model