Merck
EP5804752007
Eppendorf® Adapter For 100 mL Rectangular Bucket
holds standard, Vacutainer,Falcon and micro test tubes 2.6-7 mL, pack of 2 ea
About This Item
UNSPSC-Code:
41103911
NACRES:
NB.82
Leistungsmerkmale
holds standard, Vacutainer,Falcon and micro test tubes 2.6-7 mL
Verpackung
pack of 2 ea
Hersteller/Markenname
Eppendorf® 5804752007
Methode(n)
centrifugation: suitable
Kompatibilität
for use with 100 mL rectangular bucket
Allgemeine Beschreibung
Tube adapters for use with Eppendorf 100 mL rectangular bucket.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)