CLS6632
Corning® Step-R Repeating Pipettor Syringe Tips
volume (5.0 mL), sterile
Synonym(e):
pipet tips
Empfohlene Produkte
Sterilität
sterile
Verpackung
pack of 100 ea
Hersteller/Markenname
Corning 6632
Volumen
(5.0 mL)
Verwandte Kategorien
Allgemeine Beschreibung
- Ideal for viscous, dense and volatile liquides
- Minimum increments printed on the syringe itself
- Accurately deliver selected dose
- Made from high quality polypropylene and polyethylene
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.