Merck
CLS480140
Corning® LSE Adapters
for 10/15 mL tubes in 50 mL rotor, 6/cs
Synonym(e):
centrifuge rotor tube adapters, centrifuge tube adapters
About This Item
UNSPSC-Code:
41121815
NACRES:
SB.56
Verpackung
case of 6
pack of 6
Hersteller/Markenname
Corning 480140
Eignung
suitable for (for use with Corning LSE compact centrifuge)
Allgemeine Beschreibung
These are optional adapters for Corning LSE compact centrifuge rotors: CLS480136, CLS480137, CLS480138; CLS480139
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.