Merck
CLS285022
Corning® cover glasses
square, No. 11/2, W × L 22 mm × 22 mm
Synonym(e):
coverslips, glass covers, slide covers
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.16
Materialien
borosilicate glass
square
Verpackung
pack of 200 ea
case of 2000 ea
Hersteller/Markenname
Corning 2850-22
B × L
22 mm × 22 mm
Bindungsart
non-treated surface
Leistungsmerkmale und Vorteile
  • Corning® cover glasses are made from optically clear borosilicate glass.
  • Chemically resistant cover glasses are not affected by weathering or surface attachment.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)