Merck
CLS11410
Corning® closed system Erlenmeyer flask
pre-assembled, capacity 250 mL, sterile
Synonym(e):
conical flasks
About This Item
UNSPSC-Code:
41122104
NACRES:
NB.43
Materialien
flat bottom flask
polycarbonate
polypropylene cap
vent cap
Sterilität
sterile
Leistungsmerkmale
cap
Verpackung
pack of 4 ea
Hersteller/Markenname
Corning 11410
Kapazität
250 mL
Sonstige Hinweise
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.