Merck
Z755508
Stacking platform with flat mat for BenchRocker variable 2D platform rocker
pkg of 1 ea
About This Item
UNSPSC-Code:
41103811
NACRES:
NB.73
Verpackung
pkg of 1 ea
Hersteller/Markenname
Benchmark Scientific
Allgemeine Beschreibung
Stacking Platform, extra large 14x12 inches with flat mat and alowing 3.0 inch separation of platforms.
Rechtliche Hinweise
BenchRocker is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)