Merck
Z693758
LabJaws all-position clamp holder
nickel plated
Synonym(e):
clamp holders, frame connectors
About This Item
UNSPSC-Code:
41122413
NACRES:
NB.35
Materialien
nickel plated
Hersteller/Markenname
Troemner 916085
Backenkapazität
0-19 mm
Allgemeine Beschreibung
Permits adjustment at any angle in any plane. Holders are set at 90° to each other, connected by a 90° connector, allowing 360° rotation.
Rechtliche Hinweise
LabJaws is a trademark of Henry Troemner LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)