Z680850
Duran® stirred reactor caps and accessories
spare screw cap, 2-ports
Synonym(e):
1200390
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
Empfohlene Produkte
Hersteller/Markenname
DURAN®
Schott
Fitting
for for GL 45 stirred reactor systems
Allgemeine Beschreibung
These components allow for extra media bottles to be connected or lets sterile pressure compensation be possible. The parts are autoclaveable and the drive is provided by a standard magnetic stirrer.
Rechtliche Hinweise
DURAN is a registered trademark of DWK Life Sciences
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)