Merck
Z679127
Anti-static unisex laboratory coats
size M
Synonym(e):
laboratory apparel
About This Item
UNSPSC-Code:
46181532
NACRES:
NB.35
Größe
M
Leistungsmerkmale
CE compliant
Verfügbarkeit
not available in USA and Canada
Verwandte Kategorien
Allgemeine Beschreibung
Tough white 210 g/m cotton/polyester reusable laboratory coats, full-length with a Nega-stat grid offering electrostatic protection.
  • Concealed stud-fastening front
  • Two hip pockets and one breast pocket, all reinforced with bar tacks
  • Finished length 100 cm, with center back vent
Conforms to prEN1149-3:2001
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)