Merck
Z404942
IKA® IKATRON PT 1000 Temperaturmessfühler aus Edelstahl
standard temperature probe, 260 x 3 mm, 1/cs
Synonym(e):
contact thermometer, temperature probe
About This Item
UNSPSC-Code:
41112303
NACRES:
NB.76
Materialien
stainless steel
Verpackung
case of 1
pack of 1
Hersteller/Markenname
IKA 2735451
L × Durchm.
260 mm × 3 mm , Depth of immersion: 230 mm
Allgemeine Beschreibung
Replacement stainles steel sensor for use with IKA stirrers ETS-D5 and ETS-D6.
Rechtliche Hinweise
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
IKATRON is a trademark of IKA-Werke GmbH & Co. KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)