Merck
Z301116
Quickfit® Erweiterungsadapter
top female joint: ST/NS 24/29, bottom male joint: ST/NS 19/26
Synonym(e):
XA32
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
Verbindung
female joint: ST/NS top24/29
male joint: ST/NS bottom19/26
Hersteller/Markenname
Quickfit XA 32
Verfügbarkeit
not available in USA and Canada
not available in USA and Canada
Allgemeine Beschreibung
These adapters connect items with larger cone to a piece of glassware with a smaller size socket.
Rechtliche Hinweise
Quickfit is a registered trademark of SciLabware Limited
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)