Merck
WHA68941304
Whatman® GD/X-Spritzenfilter
glass microfiber membrane, pore size 0.45 μm, diam. 13 mm, non-sterile, pk of 150 ea
Synonym(e):
Whatman filter, Z277339, syringe filter, syringe filters
About This Item
UNSPSC-Code:
41104922
NACRES:
NB.24
Materialien
polypropylene housing
polypropylene membrane (pigment-free)
Sterilität
non-sterile
Leistungsmerkmale
glass fiber prefilter
Verpackung
pk of 150 ea
Hersteller/Markenname
Whatman 6894-1304
Whatman Article No. 28420545 (US reference)
Durchmesser
13 mm
Volumen
≤10 mL
Porengröße
0.45 μm
Fitting
female Luer inlet (lock)
male Luer outlet
Allgemeine Beschreibung
Non-sterile syringe filters with a glass membrane, suitable for hard-to-filter material with high solids. Whatman GD/X Syringe Filters are high-viscosity filters that incorporate a prefilter to enable filtration of viscous, dense or otherwise hard-to-filter material with high solids content as in environmental and food analysis samples. The prefilter contains a high-efficiency borosilicate glass filtration stack that will retain fine particles down to 0.7 μm, allowing difficult sample filtration with less hand force. It reduces the need for multiple syringe filters by processing three to seven times more sample volume than syringe filters without prefilters. The filters can be autoclaved.
Leistungsmerkmale und Vorteile
  • Non sterile Whatman GD/X Syringe Filters combine a polypropylene housing, high-efficiency Whatman GMF 150 and GF/F pre-filter media, and one of the following membrane materials:
  • Polyethersulfone (PES) for serum, plasma and tissue culture
  • Polyvinylidene difluoride (PVDF) for HPLC
  • Cellulose acetate for biological solutions
  • Borosilicate glass microfiber
Eignung
Suitable for filtering environmental and food analysis samples. Suitable for particulate laden sample preparation.
Sonstige Hinweise
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Rechtliche Hinweise
Whatman is a registered trademark of Cytiva
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)