Merck
WHA10483064
Whatman® hybridization reaction folders
Synonym(e):
Z698962
About This Item
UNSPSC-Code:
41104900
NACRES:
NB.24
Verpackung
pkg of 50 ea
Hersteller/Markenname
Whatman 10483064
Whatman Article No. 28416120 (US reference)
Allgemeine Beschreibung
Sealable hybridization bags
Whatman reaction folders are sealable bags for hybridization and incubation reactions.
The folders are open on three sides to allow easier insertion of a wet membrane when compared to standard hybridization bags. The folders can be sealed with a standard heat-sealer. Available in 8 x 9.5 inch (20.32 x 24.13 cm) size.
Leistungsmerkmale und Vorteile
  • The folders are open on three sides to allow easier insertion of a wet membrane when compared to standard hybridization bags
  • The folders can be sealed with a standard heat-sealer.
  • Available in 203.2 x 241.3 mm (8 x 9.5 in) size.
Sonstige Hinweise
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Rechtliche Hinweise
Whatman is a registered trademark of Cytiva
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)