Merck
SYNF664500
Synthware Erlenmeyer flask
capacity 500 mL, joint: ST/NS 24/40
Synonym(e):
Synthware F664500, glass conical flask
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.51
Materialien
flat bottom
glass flask
Verbindung
joint neck24/40
joint: ST/NS24/40
Sterilität
non-sterile
Leistungsmerkmale
without cap
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F664500
Kapazität
500 mL
Allgemeine Beschreibung
With joint. Stopper is not supplied. All sizes including 1L flasks are hand-blown from tubing to ensure heavy and uniform wall.
Sonstige Hinweise
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)