Merck
SYNF308100
Synthware single-neck round-bottom flask
100 mL, joint: ST/NS 29/42
Synonym(e):
Synthware F308100, glass round flask
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.51
Materialien
borosilicate glass flask
round bottom
single-neck
Verbindung
joint: ST/NS29/42
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F308100
Kapazität
100 mL
100 mL
Allgemeine Beschreibung
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)