Merck
SYNB300125C
Synthware gas washing bottle with fritted disc
125 mL, joint: ST/NS 45/50, Disc porosity C
Synonym(e):
Synthware B300125C, Drechsel bottle, gas washing bottle
About This Item
UNSPSC-Code:
41103209
NACRES:
NB.51
Materialien
clear bottle
cylindrical bottle
straight bottle
Verbindung
joint: ST/NS45/50
Leistungsmerkmale
cap
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware B300125C
Höhe
290 mm
Kapazität
125 mL
125 mL
Allgemeine Beschreibung
Gas washing bottle with standard taper 45/50 cap style stopper and fritted disc angled at 15° for even distribution of gas throughout absorbing material. Available with a coarse or extra coarse fritted disc. Order plastic clip separately. (not supplied)
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)