Merck
SLW1060/06D
Pyrex® Watch glasses
nominal diameter 50 mm
Synonym(e):
Pyrex, SciLabware 1060/06D
About This Item
UNSPSC-Code:
41121801
NACRES:
NB.43
Verpackung
pkg of 10
Hersteller/Markenname
1060/06D
SciLabware
Nenndurchmesser
50 mm
Allgemeine Beschreibung
Watch glasses
  • For evaporation work and covering heated beakers
  • Manufactured from heat resisting Pyrex borosilicate glass
  • Fire-polished edges to reduce chipping and breakage
Leistungsmerkmale und Vorteile
• Complies with DIN 12341• Manufactured from borosilicate glass 3.3• Heat resistant• Fire-polished edges to reduce chipping and breakage
Rechtliche Hinweise
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)