Merck
MT51343204
Mettler Toledo InLab® Redox Au ORP electrode
Redox Electrode with Gold Ring Indicator
About This Item
UNSPSC-Code:
39121436
NACRES:
NB.79
Materialien
glass shaft
Beschreibung
S7 connection
Without cable
Hersteller/Markenname
Mettler-Toledo 51343204
Temperaturmessbereich
0-100 °C
L × B
120 mm × 12 mm
Allgemeine Beschreibung
Mettler Toledo InLab® Redox Au is a combination electrode for measuring the oxidation/reduction power of a solution. Consists of a gold ring indicator electrode and a silver/silver chloride reference electrode.
The measuring signal is produced on the surface of the precious metal by an exchange of electrons with the oxidation-reduction system of the measured medium.
The combination design avoids use of separate reference and indicating electrodes.
The InLab® Redox Au electrode is fitted with the innovative Safelock closing ring which gives easy access for re-filling reference electrolyte and complete sealing when closed.
Different cables can be connected to the S7 screw head allowing use with a wide range of meters.
  • Combination design
  • Safelock closing ring
  • Detachable cable
Rechtliche Hinweise
InLab is a registered trademark of Mettler-Toledo, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)