Merck
DWK218062454
DURAN® graduated amber laboratory bottle
with cap, round amber glass bottle, bottle capacity (100 mL), non-sterile
Synonym(e):
218062454, DURAN® Amber Bottle, Graduated Glass Bottle
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
Materialien
round amber glass bottle
Sterilität
non-sterile
Leistungsmerkmale
with cap
Verpackung
case of 10 ea
Hersteller/Markenname
DURAN® 218062454
Flaschenkapazität
(100 mL)
Allgemeine Beschreibung
Standardised GL-thread and matching cap system for practicularly tight sealing and simple, clean pouring out. The only external application of the colour ensures that internal surfaces have standard DURAN glass properties.
Very durable and chemically resistant amber colour. Easy-to-read scale with graduation marks to estimate the content. Fired-on, highly durable white ceramic print. Sizes 10 and 25 ml with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required.
Rechtliche Hinweise
DURAN is a registered trademark of DWK Life Sciences
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)