Merck
BR723014
BRAND® Haemacytometer cover glass, for counting chambers
borosilicate glass, size 24 mm × 24 mm × 0.4 mm
Synonym(e):
Cover glasses, Microscopy
About This Item
UNSPSC-Code:
41122600
NACRES:
NB.22
Materialien
borosilicate glass
Leistungsmerkmale
IVD
Hersteller/Markenname
BRAND 723014
Größe
24 mm × 24 mm × 0.4 mm
Bindungsart
non-treated surface
Allgemeine Beschreibung
Borosilicate glass, DIN ISO 8255. Refractive index ne = 1.52 ± 0.01, Abbe number ve = 56.5 ± 0.5. Flatness tolerance ± 3 μm.
They differ from ordinary cover glasses by their plain ground and polished surface.
Verpackung
2 cover glasses in a plastic bag, separated by tissue paper.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)