Merck
BR43328
BRAND® BLAUBRAND® density bottle, Gay-Lussac pattern, calibrated
capacity 50 mL, borosilicate glass
Synonym(e):
BLAUBRAND®, Gay-Lussac
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.71
Materialien
borosilicate glass
glass stopper
narrow-mouth bottle
round bottle (Gay-Lussac)
white bottle
Verbindung
joint ST/NS neck10/19
Leistungsmerkmale
with cap
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 43328
Kapazität
50 mL
Allgemeine Beschreibung
DIN ISO 3507, type Gay-Lussac. Calibrated to contain (TC, In). Incl. individual certificate. Stopper NS 10/19 with capillary. Top of stopper ground and polished. The volume in cm³ is specified to a precision of 3 decimal places.
Rechtliche Hinweise
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)