Merck
UHPLC Analysis of Boswellic Acids on Titan C18
UHPLC Analysis of Boswellic Acids on Titan C18 application for UHPLC
Materials
mobile phase component
Product No.
Description
Pricing
Phosphoric acid
≥85 wt. % in H2O, ≥99.999% trace metals basis
standard
Product No.
Description
Pricing
α-Boswellic acid
analytical standard
β-Boswellic acid
analytical standard
11-Keto-β-boswellic acid
analytical standard
3-O-Acetyl-11-keto-β-boswellic acid
analytical standard
CONDITIONS
column
Titan C18, 10 cm x 2.1 mm I.D., 1.9 μm particles (577124-U)
mobile phase
[A] 0.1% phosphoric acid; [B] acetonitrile; (10:90, A:B)
flow rate
0.4 mL/min
pressure
2450 psi (169 bar)
column temp.
35 °C
detector
UV, 210 nm
injection
2 μL
sample
6 Boswellic Acids 50μg/mL in 70:30, water:methanol
Description
Legal Information
null