Merck
HPLC Analysis of Water-Soluble B-Vitamins on Ascentis® Express F5
HPLC Analysis of Water-Soluble B-Vitamins on Ascentis® Express F5 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express F5, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 5 cm × 3 mm
standard
Product No.
Description
Pricing
Pyridoxine hydrochloride
≥98% (HPLC)
Calcium pantothenate
meets USP testing specifications, monograph mol wt. 476.53 (C18H32CaN2O10)
Thiamine hydrochloride
reagent grade, ≥99% (HPLC)
Nicotinic acid
≥99.5% (HPLC)
Nicotinamide
≥98.5% (HPLC)
Biotin
≥99.0% (T)
Cyanocobalamin (B12)
analytical standard
Riboflavin (B2)
analytical standard
CONDITIONS
column
Ascentis Express F5, 5 cm x 3.0 mm I.D., 2.7 μm particles (53576-U)
mobile phase
[A] 20 mM potassium phosphate, dibasic, pH 6.2; [B] acetonitrile
gradient
0% B for 1.5 min, 0-10% over 0.5 min; to 15% B in 1.5 min, 2 min re-equilibration at 0% B
flow rate
1.5 mL/min
pressure
4931 psi (340 bar)
column temp.
35 °C
detector
UV, 210 nm
injection
5 μL
sample
all vitamins in water, 10 μg/mL for nicotinic acid and folic acid, 12 μg/mL for thiamine hydrochloride and cyanocobalamin, (20 μg/mL for pyridoxine, 30 μg/mL for riboflavin, 40 μg/mL for niacineamide, 100 μg/mL for calcium pantothenate, 120 μg/mL for biotin)
Description
Analysis Note
A mix of 9 water-soluble B-vitamins was separated on Ascentis Express F5 column
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany