HPLC Analysis of Sulconazole Enantiomers on Astec® Cellulose DMP
![HPLC Analysis of Sulconazole Enantiomers on Astec® Cellulose DMP application for HPLC](/deepweb/assets/sigmaaldrich/product/images/362/393/b0cb0b41-8ed3-48c9-ab3d-b08774a8f925/800/b0cb0b41-8ed3-48c9-ab3d-b08774a8f925.jpg)
CONDITIONS
column
Astec Cellulose DMP, 25 cm x 4.6 mm I.D., 5 μm particles (51099AST)
mobile phase
[A] heptane; [B] isopropanol; (80:20, A:B)
flow rate
1 mL/min
column temp.
30 °C
detector
UV, 254 nm
injection
15 μL
sample
1 g/L in 50:50, heptane:isopropanol