HPLC Analysis of Proteins on Antibodix® WCX-NP10, Comparison of WCX Columns
![HPLC Analysis of Proteins on Antibodix® WCX-NP10, Comparison of WCX Columns application for HPLC](/deepweb/assets/sigmaaldrich/product/images/499/275/d38c34b0-7174-4df7-a15c-8ca2c94edd79/800/d38c34b0-7174-4df7-a15c-8ca2c94edd79.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Antibodix WCX-NP10, 25 cm x 4.6 mm I.D., 10 μm particles (Z777272)
mobile phase
(A) 10 mM sodium phosphate buffer, pH 6.0; (B) A + 1.0 M NaCl
gradient
0 to 100% B in 42 min
flow rate
1.0 mL/min
column temp.
ambient
detector
UV, 214 nm
injection
5 μL
sample
1 mg/mL each
Description
Legal Information
Antibodix is a registered trademark of Sepax Technologies