HPLC Analysis of Peptides on Discovery® BIO Wide Pore C18, Preparative Scale
![HPLC Analysis of Peptides on Discovery® BIO Wide Pore C18, Preparative Scale application for HPLC](/deepweb/assets/sigmaaldrich/product/images/135/578/1ca6c0d7-d994-4ed8-896c-2d8772a57aaf/800/1ca6c0d7-d994-4ed8-896c-2d8772a57aaf.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery BIO Wide Pore C18, 15 cm × 10 mm I.D., 10 μm particles (567208-U)
mobile phase
[A] (80:20), 0.1% TFA in water:0.1% TFA in acetonitrile; [B] (66:34), 0.1% TFA in water:0.1% TFA in acetonitrile
flow rate
4.7 mL/min
column temp.
30 °C
detector
UV, 215 nm
injection
24.5 μL
sample
Cat. No. P2693
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany