Merck
HPLC Analysis of Ketamine Enantiomers on Astec® Cellulose DMP
HPLC Analysis of Ketamine Enantiomers on Astec® Cellulose DMP application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® Cellulose DMP Chiral HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
standard
Product No.
Description
Pricing
S-(+)-Ketamine hydrochloride
(±)-Ketamine hydrochloride
solid
Ketamine hydrochloride/xylazine hydrochloride solution
CONDITIONS
column
Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 μm particles (51098AST)
mobile phase
[A] heptane; [B] IPA; [C] DEA; (90:10:0.1, A:B:C)
flow rate
0.5 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
2 μL
sample
2 mg/mL in mobile phase