HPLC Analysis of Glutamic Acid Enantiomers on Astec® CHIROBIOTIC® T (25 °C)
![HPLC Analysis of Glutamic Acid Enantiomers on Astec® CHIROBIOTIC® T (25 °C) application for HPLC](/deepweb/assets/sigmaaldrich/product/images/673/368/8daaee3c-415b-403d-b355-8e00b53f739e/800/8daaee3c-415b-403d-b355-8e00b53f739e.jpg)
Materials
analytical column
Product No.
Description
Pricing
guard cartridge
Product No.
Description
Pricing
Astec® CHIROBIOTIC® T Chiral HPLC Guard Column
Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mmstandard
Product No.
Description
Pricing
L-Glutamic acid
from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%L-Glutamic acid
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, SwitzerlandCONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] water; [B] methanol; [C] formic acid; (30:70:0.02, A:B:C)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 205 nm
injection
10 μL
sample
300 μg /mL in 30:70 water:methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC