HPLC Analysis of DNA on Proteomix® WAX-NP5
![HPLC Analysis of DNA on Proteomix® WAX-NP5 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/407/186/c31b8240-c491-4e97-ac6e-8f187d55ee18/800/c31b8240-c491-4e97-ac6e-8f187d55ee18.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Proteomix WAX-NP5, 15 cm x 4.6 mm I.D., 5 μm particles (Z777262)
mobile phase
(A) 20 mmol Tris, pH 6.0; (B) A + 1.0 M NaCl
gradient
20 to 100% B in 40 min
flow rate
0.6 mL/min
pressure
1015 psi (70 bar)
temp.
ambient
detector
UV, 260 nm
injection
5 μL
sample
DNA
Description
Legal Information
Proteomix is a registered trademark of Sepax Technologies