HPLC Analysis of Diazepam and Metabolites in Serum on Discovery® C18 after SPE using Discovery® DSC-8
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
sample preparation
SPE (Solid Phase Extraction)
sample/matrix
goat serum spiked with diazepam and metabolites at 0.5 μg/mL and diluted 1:1 in 25 mM ammonium formate, pH 7.1
SPE well plate
Discovery DSC-8 96-well SPE Plate, 100 mg/well (575627-U)
condition
1 mL methanol followed by 1 mL DI water.
sample addition
1 mL spiked goat serum
washing
1 mL 25% methanol in 25 mM ammonium formate, pH 2.75
elution
1 mL 60% methanol in 25 mM ammonium formate, pH 2.75
elution
Evaporate eluate with nitrogen purge (30 °C; ~15 min.). Reconstitute in 200 μL mobile phase.
column
Discovery C18, 5 cm x 4.6 mm I.D., 5 μm particles (504947)
mobile phase
[A] Methanol; [B]10 mM ammonium acetate, pH 4.5;(45:55, A:B)
flow rate
1.5 mL/min
column temp.
35 °C
detector
UV, 240 nm
injection
25 μL
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany