Merck
  • Lithium secondary batteries using modified-imidazolium room-temperature ionic liquid.
Lithium secondary batteries using modified-imidazolium room-temperature ionic liquid.
The journal of physical chemistry. B (2006-05-26)
Shiro Seki, Yo Kobayashi, Hajime Miyashiro, Yasutaka Ohno, Akira Usami, Yuichi Mita, Nobuo Kihira, Masayoshi Watanabe, Nobuyuki Terada
ABSTRACT
Highly reversible, safe lithium secondary batteries that use imidazolium-cation-based room-temperature ionic liquid as an electrolyte and lithium metal as an anode material were realized by the molecular design. To achieve higher reduction stability, an electron-donating substituent was introduced to promote charge delocalization in the imidazolium cation of room-temperature ionic liquids.