MilliporeSigma
  • Efficient aldolase catalytic antibodies that use the enamine mechanism of natural enzymes.
Efficient aldolase catalytic antibodies that use the enamine mechanism of natural enzymes.
Science (New York, N.Y.) (1995-12-15)
J Wagner, R A Lerner, C F Barbas
ABSTRACT
Antibodies that catalyze the aldol reaction, a basic carbon-carbon bond-forming reaction, have been generated. The mechanism for antibody catalysis of this reaction mimics that used by natural class I aldolase enzymes. Immunization with a reactive compound covalently trapped a Lys residue in the binding pocket of the antibody by formation of a stable vinylogous amide. The reaction mechanism for the formation of the covalent antibody-hapten complex was recruited to catalyze the aldol reaction. The antibodies use the epsilon-amino group of Lys to form an enamine with ketone substrates and use this enamine as a nascent carbon nucleophile to attack the second substrate, an aldehyde, to form a new carbon-carbon bond. The antibodies control the diastereofacial selectivity of the reaction in both Cram-Felkin and anti-Cram-Felkin directions.