Merck
  • Use of reversed phase HP liquid chromatography to assay conversion of N-acylglycines to primary fatty acid amides by peptidylglycine-alpha-amidating monooxygenase.
Use of reversed phase HP liquid chromatography to assay conversion of N-acylglycines to primary fatty acid amides by peptidylglycine-alpha-amidating monooxygenase.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2004-07-30)
Tara Carpenter, Derek D Poore, Andrew J Gee, Pallavi Deshpande, David J Merkler, Mitchell E Johnson
ABSTRACT
Primary fatty acid amides (R-CO-NH2) and N-acylglycines (R-CO-NH-CH2-COOH) are classes of compounds that have only recently been isolated and characterized from biological sources. Key questions remain regarding how these lipid amides are produced and degraded in biological systems. Relative to the fatty acids, little has been done to develop methods to separate and quantify the fatty acid amides and N-acylglycines. We describe reversed phase HPLC methods for the separation of C2-C12 primary fatty acid amides and N-acylglycines and also C12-C22 fatty acid amides. Separation within each class occurs primarily on the basis of simple interactions between the acyl chain and the chromatographic stationary phase, but the polar headgroups on these and related fatty acids and N-acylethanolamides modulate the absolute retention in reversed phase mode. We use these methods to measure the enzyme-mediated, two-step conversion of N-octanoylglycine to octanoamide.