Merck
  • Modeling the sorption kinetic of metsulfuron-methyl on Andisols and Ultisols volcanic ash-derived soils: kinetics parameters and solute transport mechanisms.
Modeling the sorption kinetic of metsulfuron-methyl on Andisols and Ultisols volcanic ash-derived soils: kinetics parameters and solute transport mechanisms.
Journal of hazardous materials (2010-04-20)
Lizethly Cáceres, Mauricio Escudey, Edwar Fuentes, María E Báez
ABSTRACT
Metsulfuron-methyl sorption kinetic was studied in Andisol and Ultisol soils in view of their distinctive physical and chemical properties: acidic pH and variable surface charge. Different kinetic models were applied to the experimental results. The pseudo-second-order model fitted sorption kinetics data better than the pseudo-first-order model. The rate constant and the initial rate constant values obtained through this model demonstrated the different behavior of metsulfuron-methyl in both kinds of soils, both parameters being the highest for Andisol. The application of Elovich equation, intraparticle diffusion model and a two-site nonequilibrium model (TSNE) allowed to conclude that: (i) the high organic matter content is the governing factor for Andisols where mass transfer across the boundary layer, and in a lesser degree, intraparticle diffusion were the two processes controlling sorption kinetic and (ii) the mineral composition was more relevant in Ultisols where rate was controlled almost exclusively by intraparticle diffusion into macropores and micropores. The slower sorption rate on Ultisols, the mechanism involved and the lower sorption capacity of this kind of soils must be taken into account to assess leaching behavior of this herbicide.
Product Description
Supelco
Metsulfuron-methyl, PESTANAL®, analytical standard