Merck
  • Comparison of the maternal and neonatal effects of bupivacaine plus fentanyl and ropivacaine plus fentanyl during cesarean delivery.
Comparison of the maternal and neonatal effects of bupivacaine plus fentanyl and ropivacaine plus fentanyl during cesarean delivery.
Nigerian journal of clinical practice (2013-04-09)
Un Canan, D Ornek, O Kilci, F Donmez, M Gamli, B Dikmen
ABSTRACT
The aim of the present study was to compare the anesthetic efficacy, and fetal and maternal effects of 7.5 mg (1 ml) intrathecal 0.75% hyperbaric ropivacaine + 25 μg (0.5 ml) fentanyl versus 5 mg (l ml) intrathecal 0.5% hyperbaric bupivacaine + 25 μg (0.5 ml) fentanyl in elective cesarean delivery. The study included 40 ASA I-II cases scheduled for cesarean delivery that were randomized into two groups of 20 cases each. Cases in the RF group were administered 0.75% hyperbaric ropivacaine + 25 μg (0.5 ml) fentanyl and those in the BF group were administered 5 mg (l ml) hyperbaric bupivacaine + 25 μg (0.5 ml) fentanyl into the spinal space. The time until spinal anesthesia in the T4 dermatome, overall duration of analgesia, hemodynamic parameters, Apgar score of newborns at 1-5 min, fetal blood gas values (pH, PO2, PCO2, HCO3-, and BE), maternal side effects, the degree of motor block, maternal need for ephedrine, objective pain scale score, and patient satisfaction were recorded in each group. There were no significant differences between the groups in terms of the parameters evaluated (P > 0.05). In elective cesarean delivery, the combinations of bupivacaine + fentanyl or ropivacaine + fentanyl exhibited similar anesthetic efficacy, and fetal and maternal effects.
Product Description
Supelco
Bupivacaine hydrochloride monohydrate, analytical standard, for drug analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Fentanyl citrate salt
SKU
Pack Size
Availability
Price
Quantity
Supelco
Fentanyl solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
SKU
Pack Size
Availability
Price
Quantity
Supelco
Fentanyl solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
SKU
Pack Size
Availability
Price
Quantity
Supelco
Bupivacaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material