Skip to Content
Merck
All Photos(2)

Documents

V900149

Sigma-Aldrich

Ammonium tartrate dibasic

Vetec, reagent grade, 99%

Synonym(s):

L-(+)-Tartaric acid diammonium salt, Diammonium tartrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2C4H4O6
CAS Number:
Molecular Weight:
184.15
Beilstein:
6120352
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

99%

form

crystals

pH

5-7

SMILES string

N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O

InChI

1S/C4H6O6.2H3N/c5-1(3(7)8)2(6)4(9)10;;/h1-2,5-6H,(H,7,8)(H,9,10);2*1H3/t1-,2-;;/m1../s1

InChI key

NGPGDYLVALNKEG-OLXYHTOASA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carles Cruz-Morató et al.
The Science of the total environment, 493, 365-376 (2014-06-22)
Hospital effluents contribute to the occurrence of emerging contaminants in the environment due to their high load of pharmaceutical active compounds (PhACs) and some endocrine disruptor compounds (EDCs). Nowadays, hospital wastewaters are co-treated with urban wastewater; however, the dilution factor
Laura Ferrando-Climent et al.
Chemosphere, 136, 9-19 (2015-04-26)
This work presents a study about the elimination of anticancer drugs, a group of pollutants considered recalcitrant during conventional activated sludge wastewater treatment, using a biological treatment based on the fungus Trametes versicolor. A 10-L fluidized bed bioreactor inoculated with
Hongquan Wan et al.
Nature communications, 6, 6114-6114 (2015-02-11)
A(H1N1)pdm09 influenza A viruses predominated in the 2013-2014 USA influenza season, and although most of these viruses remain sensitive to Food and Drug Administration-approved neuraminidase (NA) inhibitors, alternative therapies are needed. Here we show that monoclonal antibody CD6, selected for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service