Skip to Content
Merck
All Photos(2)

Documents

V800339

Sigma-Aldrich

Potassium sulfate

LR, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2SO4
CAS Number:
Molecular Weight:
174.26
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

LR

product line

Vetec

Assay

≥99%

form

crystalline

pH

5.5-8.5 (5% in water)

SMILES string

[K+].[K+].[O-]S([O-])(=O)=O

InChI

1S/2K.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

OTYBMLCTZGSZBG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinjian Shi et al.
Nature communications, 5, 4775-4775 (2014-09-03)
Tungsten trioxide/bismuth vanadate heterojunction is one of the best pairs for solar water splitting, but its photocurrent densities are insufficient. Here we investigate the advantages of using helical nanostructures in photoelectrochemical solar water splitting. A helical tungsten trioxide array is
Viralkumar Patel et al.
The American journal of gastroenterology, 104(4), 953-965 (2009-02-26)
Ingestion of a concentrated low-volume phosphate solution produces copious diarrhea, which cleanses the colon, but it occasionally causes renal failure due to calcium phosphate precipitation in renal tubules. We hypothesized that a concentrated low-volume sulfate solution would be an equally
Douglas K Rex et al.
Gastrointestinal endoscopy, 72(2), 328-336 (2010-07-22)
Low-volume bowel preparations for colonoscopy improve tolerability. We compared the efficacy, tolerability, and safety of a new low-volume sulfate solution with a standard 4-L polyethylene glycol electrolyte lavage solution as bowel preparation for colonoscopy. Multicenter, single-blind, randomized, noninferiority study. Five
Igor Moudrakovski et al.
The journal of physical chemistry. A, 114(1), 309-316 (2010-01-09)
A set of potassium sulfates presenting a variety of sulfur environments (K(2)SO(4), KHSO(4), K(2)S(2)O(7), and K(2)S(2)O(8)) has been studied by (33)S solid state NMR at 21 T. Low natural abundance (0.75%) and small gyromagnetic ratio of (33)S presented a serious
Sunghee Lee et al.
Langmuir : the ACS journal of surfaces and colloids, 26(12), 9556-9564 (2010-02-18)
In this work, we have investigated the effect of potassium salts of different anions upon the crystal nucleation of K(2)SO(4) as interfacially templated by a surfactant monolayer of 1-octadecylamine (ODA), in an aqueous microdroplet system bounded by a liquid-liquid interface

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service