G005896
HPLC Analysis of a Monoclonal Antibody on Zenix®-C SEC-150 after Partial Proteolysis with IdeS Protease
application for HPLC
About This Item
technique(s)
HPLC: suitable
test parameters
column: Zenix-C SEC-150, 30 cm X 4.6 mm I.D., 3 μm particles (Z777083)
mobile phase: 0.15 M potassium phosphate monobasic, pH 7.0 with potassium hydroxide (567329-U)
flow rate: 0.35 mL/min
pressure: 1450 psi (100 bar)
column temp.: ambient
detector: UV, 215 nm
injection: 1 μL
sample: digested SO57 mAb, ~0.5 g/L
suitability
application for HPLC
application(s)
life science and biopharma
Analysis Note
Legal Information
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service