Key Documents
57695
Rheodyne® Model 9010 Injector
About This Item
UNSPSC Code:
41115854
Pricing and availability is not currently available.
feature
manual
number of ports 6
positions 2
parameter
5000 psi max. pressure
General description
This injector is similar in function to the Model 7010 stainless steel injector, but is constructed of inert PEEK and Tefzel materials for use in the pH range 0-14. It can be used for HPLC, soft gel chromatography, or ion chromatography.
A position-sensing switch closes in the INJECT position and stays closed until the handle is returned to the LOAD position.
Supplied with a 20 μL sample loop; order additional loops separately.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Rheodyne is a registered trademark of IDEX Health & Science, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)