Skip to Content
Merck
All Photos(1)

Documents

442768

Supelco

Propionaldehyde-2,4-DNPH

analytical standard

Synonym(s):

Propionaldehyde-2,4-dinitrophenylhydrazone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
238.20
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

152-155 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

CC\C=N\Nc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C9H10N4O4/c1-2-5-10-11-8-4-3-7(12(14)15)6-9(8)13(16)17/h3-6,11H,2H2,1H3/b10-5+

InChI key

NFQHZOZOFGDSIN-BJMVGYQFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service