Key Documents
Z741674
Sartorius VALUEPAK pipette tips
volume range 50-1,200 μL
About This Item
UNSPSC Code:
41121600
maximum volume:
1.2 mL
volume range:
50-1,200 μL
sterility:
non-sterile
feature:
DNase free
RNase free
barrier: no
endotoxin free
packaging:
pack of 960 ea (refil)
pkg of refill
manufacturer/tradename:
Sartorius 791202MAX
Pricing and availability is not currently available.
material
colorless
sterility
non-sterile
feature
DNase free
RNase free
barrier: no
endotoxin free
packaging
pack of 960 ea (refil)
pkg of refill
manufacturer/tradename
Sartorius 791202MAX
maximum volume
1.2 mL
volume range
50-1,200 μL
General description
Large value packs of tips with the perfect tip fitting and sealing to the pipetter. This gives you the best possible accuracy and precision at the best possible price. Compatible with Optiload feature in Biohit Picus, eLINE®, mLINE® and Proline® Plus pipetters, enabling ergonomic and light tip attachment and ejection.
Other Notes
Browse all pipette tips
Legal Information
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
eLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
Choose from one of the most recent versions:
Certificates of Analysis (COA)