Key Documents
Z360457
Utility boxes, plastic
L × W × H 129 mm × 75 mm × 59 mm
Synonym(s):
laboratory storage, plastic storage box
About This Item
UNSPSC Code:
41123300
NACRES:
NB.51
Pricing and availability is not currently available.
material
styrene-acrylonitrile (SAN)
manufacturer/tradename
Nalgene 5700-0325
L × W × H
129 mm × 75 mm × 59 mm
General description
Practical containers for multipurpose lab storage. Glass-clear SAN body; white LDPE lid with white SAN insert. Ideal for dry materials like powders, chromatography sheets. Rounded corners for easy cleaning; stackable for compact freezer storage. Lids snap into place, but are not leakproof.
Dimensions are L × W × H
Choose from one of the most recent versions:
Certificates of Analysis (COA)